Order Entry
Netherlands
ContactUsLinkComponent
N-Octyl-4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate ≥94.0% (by GC)
N-Octyl-4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate ≥94.0% (by GC)
Catalog # TCIAO0436-100MG
Supplier:  TCI
CAS Number:  
N-Octyl-4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate ≥94.0% (by GC)
Catalog # TCIAO0436-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (GC):
    min. 94.0 %
  • Melting point:
    53.0 to 57.0 deg-C

Specification Test Results

About this item

 :
 :472.23 g/mol
CAS Number:1160823-76-6
Formula:C₁₅H₁₇Br₂FO₂S₂
MDL Number:MFCD19940962