
9-Oxoacridine-10-acetic acid (CMA) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:38609-97-1
:
Formula:C₁₅H₁₁NO₃
MDL Number:MFCD00157050
:253.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together