Oxazolo[4,5-b]pyridin-2(3H)-one ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:60832-72-6
:
Formula:C₆H₄N₂O₂
MDL Number:MFCD00204215
:136.11 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:212.0 to 216.0 deg-C
Specification Test Results
Frequently Bought Together