Oxiracetam ≥96.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
Synonyms: 2-(4-Hydroxy-2-oxo-1-pyrrolidinyl)acetamide, 2-(4-Hydroxy-2-oxopyrrolidin-1-yl)acetamide
CAS Number:62613-82-5
:
Formula:C₆H₁₀N₂O₃
MDL Number:MFCD00242951
:158.16 g/mol
Merck Index:14,06942
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:170.0 to 174.0 deg-C
Specification Test Results
Frequently Bought Together