Octenidine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:70775-75-6
:
Formula:C₃₆H₆₂N₄
MDL Number:MFCD01938808
:623.84 g/mol
Merck Index:14,06754
EINECS:274-861-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:212.0 to 216.0 deg-C
Specification Test Results
Frequently Bought Together