5-n-Octyl-2,2'-bithiophene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:93164-73-9
:
Formula:C₁₆H₂₂S₂
:278.47 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together