Order Entry
Netherlands
ContactUsLinkComponent
 
Octadecyl-3-mercaptopropionate ≥85.0% (by GC) (contains ca. 12% Hexadecyl 3-Mercaptopropionate)
 :  TCI
 
Octadecyl-3-mercaptopropionate ≥85.0% (by GC) (contains ca. 12% Hexadecyl 3-Mercaptopropionate)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 85.0 %

 

Frequently Bought Together