Order Entry
Netherlands
ContactUsLinkComponent
2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin ≥98.0% (by HPLC)
2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin ≥98.0% (by HPLC)
Catalog # TCIAO0319-100MG
Supplier:  TCI
CAS Number:  
2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin ≥98.0% (by HPLC)
Catalog # TCIAO0319-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark purple powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 95.0 %

Specification Test Results

About this item

 :
 :1118.48 g/mol
CAS Number:121399-88-0
Formula:C₄₄H₂F₂₈N₄