Octaethylene glycol monomethyl ether ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25990-96-9
:
Formula:C₁₇H₃₆O₉
MDL Number:MFCD06797152
Boiling Pt:216°C
:384.47 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together