
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53558-93-3
:
Formula:C₅H₆O₄
MDL Number:MFCD00011567
:130.10 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together