

2,7-Octadienol (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23578-51-0
:
Formula:C₈H₁₄O
MDL Number:MFCD00191472
:126.20 g/mol
Flash Pt:83°C
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
Specification Test Results
Frequently Bought Together