cis,cis-9,12-Octadecadien-1-ol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:506-43-4
:
Formula:C₁₈H₃₄O
MDL Number:MFCD00056667
:266.47 g/mol
Boiling Pt:155°C
EINECS:208-038-1
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together