Order Entry
Netherlands
ContactUsLinkComponent
 
Flammable
Health
1,4-Octadiene (cis and trans mixture) cis- and trans-mixture
Catalog # TCIAO0170-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
1,4-Octadiene (cis and trans mixture) cis- and trans-mixture
Catalog # TCIAO0170-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomers)

Specification Test Results

About this item

 :
 :110.20 g/mol
EINECS:227-132-3
UN:2309
ADR:3,II
 :GHS02
 :GHS08
CAS Number:5675-25-2
Formula:C₈H₁₄
MDL Number:MFCD00027235