
N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide ≥98.0% (by total nitrogen basis)
Catalog # TCIAO0167-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid to slightly cloudy liquid
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:275.39 g/mol
Boiling Pt:158°C
Flash Pt:177°C
EINECS:204-029-1
UN:2810
ADR:6.1,III
:GHS06
CAS Number:113-48-4
Formula:C₁₇H₂₅NO₂
MDL Number:MFCD00072467