Order Entry
Netherlands
ContactUsLinkComponent
Oxamic hydrazide ≥95.0% (by titrimetric analysis)
Flammable
Oxamic hydrazide ≥95.0% (by titrimetric analysis)
Catalog # TCIAO0156-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Oxamic hydrazide ≥95.0% (by titrimetric analysis)
Catalog # TCIAO0156-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Bromination Method):
    min. 95.0 %

Specification Test Results

About this item

 :
 :103.08 g/mol
Merck Index:14,08445
EINECS:208-213-2
UN:1325
ADR:4.1,III
 :GHS02
CAS Number:515-96-8
Formula:C₂H₅N₃O₂
MDL Number:MFCD00008004