9-(1-Naphthyl)-10-(2-naphthyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:855828-36-3
:
Formula:C₃₄H₂₂
MDL Number:MFCD20040456
:430.55 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together