
4-Nitrophenyl phosphate solution for ELISA, ready-to-use
Catalog # TCIAN1109-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Alkaline Phosphatase detection test:to pass test
Specification Test Results
About this item
:
:219.09 g/mol
:GHS08
Storage Temperature:Freezer
CAS Number:330-13-2
Formula:C₆H₆NO₆P