Order Entry
Netherlands
ContactUsLinkComponent
Diamond nanoparticles 5% dispersion in water (Carboxyl-modified) (<10 nm)
Diamond nanoparticles 5% dispersion in water (Carboxyl-modified) (<10 nm)
Catalog # TCIAN1084-5G
Leverancier:  TCI
CAS-nummer:  
Diamond nanoparticles 5% dispersion in water (Carboxyl-modified) (<10 nm)
Catalog # TCIAN1084-5G
Leverancier:  TCI
CAS-nummer:  

Specificaties

  • Pk:
    5 g
  • Seq:
    3173

Specificaties

Specificatie testresultaten

  • Appearance:
    Light gray to Brown to Black clear liquid to cloudy liquid
  • Concentration:
    4.5 to 5.4 %

Specificatie testresultaten

Over dit artikel

 :
CAS-nummer:7782-40-3
MDL-nummer:MFCD00211867