
2,7-Naphthalenebis(trifluoromethanesulphonate) ≥98.0% (by GC)
: TCI
:151391-00-3
:
:C₁₂H₆F₆O₆S₂
:MFCD20260329
:424.28 g/mol
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
Frequently Bought Together