
(4-Nitrophenyl)(phenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:905718-45-8
:
Formula:C₁₃H₉F₃INO₅S
MDL Number:MFCD21608480
:475.18 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 97.0 %
- Melting point:198.0 to 202.0 deg-C
Specification Test Results
Frequently Bought Together