

3-Nitro-2,6-lutidine ≥98.0% (by GC, titration analysis)
: TCI
:15513-52-7
:
:C₇H₈N₂O₂
:MFCD00023503
:152.15 g/mol
:105°C
:37°C
:Refrigerator
:239-543-5
:1325
:4.1,III
:GHS02
:GHS07
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:36.0 to 40.0 deg-C
Frequently Bought Together