
1-(4-Nitrophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAN0995-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:197.22 g/mol
EINECS:223-021-9
UN:2811
ADR:6.1,III
:GHS06
CAS Number:3696-22-8
Formula:(C₆H₄(NO₂))NC(S)NH₂
MDL Number:MFCD00007304