Niceritrol ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:5868-05-3
:
Formula:C₂₉H₂₄N₄O₈
MDL Number:MFCD00215982
Merck Index:14,06497
EINECS:227-519-7
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):9.50 to 10.50 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
Frequently Bought Together