
Noscapine hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAN0963-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+43.0 to +47.0 deg (C=0.5, 0.1mol/L HCl) (calcd.on anh.substance)
- Water:2.0 to 6.0 %
Specification Test Results
About this item
:
:449.88 g/mol
EINECS:213-014-9
:GHS07
Storage Temperature:Refrigerator
CAS Number:912-60-7
Formula:C₂₂H₂₃NO₇·HCl·xH₂O
MDL Number:MFCD00149223