
4-(2-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:918655-03-5
:
Formule:C₁₆H₁₃BO₂
MDL-nummer:MFCD09260454
:248.0800 g/mol
:GHS07
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 108.0 %
Specificatie testresultaten
Gerelateerde informatie
Frequently Bought Together