N-(4-Nitrobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59642-21-6
:
Formula:C₁₀H₁₀N₂O₅
MDL Number:MFCD00009794
:238.20 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:163.0 to 167.0 deg-C
Specification Test Results
Frequently Bought Together