
3-Nitro-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:26621-44-3
:
Formula:C₃H₃N₃O₂
MDL Number:MFCD00159621
:113.08 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:174.0 to 178.0 deg-C
Specification Test Results
Frequently Bought Together