
1,5-Naphthalenebis(trifluoromethanesulphonate) ≥98.0% (by GC)
Catalog # TCIAN0925-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C
Specification Test Results
About this item
:
:424.28 g/mol
:GHS07
CAS Number:152873-79-5
Formula:C₁₂H₆F₆O₆S₂
MDL Number:MFCD23380211