Nimodipine ≥98.0% (by HPLC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:66085-59-4
:
Formule:C₂₁H₂₆N₂O₇
MDL-nummer:MFCD00153848
:418.45 g/mol
Merck index:14,06551
EINECS:266-127-0
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):6.55 to 6.80 %
- Melting point:124.0 to 128.0 deg-C
Specificatie testresultaten
Frequently Bought Together