4-Nitrophenyl-β-D-cellobioside ≥98.0% (by HPLC)
TCIAN0867-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:-80.0 to -84.0 deg (C=1, methanol)
:
:463.39 g/mol
:245°C
:Freezer
:3482-57-3
:C₁₈H₂₅NO₁₃
:MFCD00069845