Order Entry
Netherlands
ContactUsLinkComponent
Neu5Ac-α(2-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Neu5Ac-α(2-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAN0860-5MG
Supplier:  TCI
CAS Number:  
Neu5Ac-α(2-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAN0860-5MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystaline
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

About this item

 :
 :754.65 g/mol
Storage Temperature:Freezer
CAS Number:2149582-14-7
Formula:C₂₉H₄₂N₂O₂₁