Order Entry
Netherlands
ContactUsLinkComponent
Neu5Ac-α(2-6)Gal-β(1-3)GlcNAc-β-pNP ≥97.0% (by HPLC)
Neu5Ac-α(2-6)Gal-β(1-3)GlcNAc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAN0855-1MG
Supplier:  TCI
CAS Number:  
undefined
Neu5Ac-α(2-6)Gal-β(1-3)GlcNAc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAN0855-1MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

About this item

 :
 :795.70 g/mol
Storage Temperature:Freezer
Formula:C₃₁H₄₅N₃O₂₁