
6-(1-Naphthylamino)-1,4-benzodioxane ≥97.0% (by GC)
Catalog # TCIAN0833-1G
Supplier: TCI
CAS Number:
Specifications
- VPE:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:133.0 to 137.0 deg-C
Specification Test Results
About this item
:
:277.32 g/mol
:GHS07
CAS Number:1820711-54-3
Formula:C₁₈H₁₅NO₂
MDL Number:MFCD11975435