(R)-(+)-1-(2-Naphthyl)ethanol ≥98.0% (by GC)
Catalog # TCIAN0784-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
- Specific rotation [a]20/D:+37.0 to +42.0 deg (C=1, methanol)
Specification Test Results
About this item
:
:172.23 g/mol
CAS Number:52193-85-8
Formula:C₁₂H₁₂O
MDL Number:MFCD00075344