
5-Norbornene-2,3-dimethanol predominantly endo isomer ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:85-39-2
:
Formule:C₉H₁₄O₂
MDL-nummer:MFCD00474435
:154.21 g/mol
EINECS:201-601-2
:GHS07
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 % (total of isomer)
- Solubility in Methanol:very faint turbidity
Specificatie testresultaten
Frequently Bought Together