Order Entry
Netherlands
ContactUsLinkComponent
Methyl-5-Norbornene-2-carboxylate (endo and exo mixture) ≥96.0% (by GC)
Irritant
Methyl-5-Norbornene-2-carboxylate (endo and exo mixture) ≥96.0% (by GC)
Catalog # TCIAN0691-25G
Supplier:  TCI
CAS Number:  
Irritant
Methyl-5-Norbornene-2-carboxylate (endo and exo mixture) ≥96.0% (by GC)
Catalog # TCIAN0691-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0% (total of isomer)

Specification Test Results

About this item

 :
 :152.19 g/mol
 :GHS07
CAS Number:6203-08-3
Formula:C₉H₁₂O₂
MDL Number:MFCD00167592