Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
 
trans,cis-2,6-Nonadien-1-ol ≥97.0% (by GC)
  TCIAN0593-5ML
 :  TCI
 :  
 
trans,cis-2,6-Nonadien-1-ol ≥97.0% (by GC)
  TCIAN0593-5ML
 :  TCI
 :  

 

  • VPE:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :140.23 g/mol
 :100°C
 :93°C
 :248-816-8
 :GHS07
 :28069-72-9
 :C₉H₁₆O
 :MFCD00014055