1,4,5,8-Naphthalenetetracarboxdiimide ≥95.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:5690-24-4
:
Formula:C₁₄H₆N₂O₄
MDL Number:MFCD00047649
:266.21 g/mol
EINECS:227-159-0
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
Frequently Bought Together