Order Entry
Netherlands
ContactUsLinkComponent
2-Nonene (cis- and trans- mixture) cis- and trans-mixture ≥94.0% (by GC)
Flammable
Health
2-Nonene (cis- and trans- mixture) cis- and trans-mixture ≥94.0% (by GC)
Supplier:  TCI
Certificates
Flammable
Health
2-Nonene (cis- and trans- mixture) cis- and trans-mixture ≥94.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 94.0% (total of isomers)

Specification Test Results

Frequently Bought Together