
Neostigmine methyl sulphate ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:51-60-5
:MFCD01558739
Formula:C₁₃H₂₂N₂O₆S
MDL Number:MFCD00011796
:334.39 g/mol
Merck Index:14,06464
EINECS:200-109-5
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together