




4-Nitrosophenol ≥98.0% (by titrimetric analysis) wetted with ca. 40% water (unit weight on dry weight basis)
TCIAN0374-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
:
:123.11 g/mol
:14,06642
:203-251-6
:1325
:4.1,II
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
:104-91-6
:C₆H₅NO₂
:MFCD00002329