
2-Nitrobenzenesulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:80-82-0
:
Formula:C₆H₅NO₅S
MDL Number:MFCD00053617
:203.17 g/mol
EINECS:201-311-6
UN:2305
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 16.0 %
Specification Test Results
Frequently Bought Together