Order Entry
Netherlands
ContactUsLinkComponent
Disodium-2,7-naphthalenedisulphonate ≥98.0% (by titrimetric analysis)
Disodium-2,7-naphthalenedisulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0014-25G
Supplier:  TCI
CAS Number:  
undefined
Disodium-2,7-naphthalenedisulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0014-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 % (calcd.on dried substance)
  • Drying loss:
    max. 6.0 %

Specification Test Results

About this item

 :
 :332.25 g/mol
EINECS:216-733-6
CAS Number:1655-35-2
Formula:C₁₀H₈O₆S₂
MDL Number:MFCD00066365