
Monomethyl succinate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3878-55-5
:
Formula:C₅H₈O₄
MDL Number:MFCD00002788
:132.11 g/mol
Boiling Pt:151°C
EINECS:223-408-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:56.0 to 60.0 deg-C
Specification Test Results
Frequently Bought Together