
2-Methoxy-6-methylbenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6161-65-5
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00463871
:166.1700 g/mol
:GHS05
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:139.0 to 143.0 deg-C
Specification Test Results
Frequently Bought Together