
2-Methoxy-4-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
TCIAM3186-1G
: TCI
:
- Pk:1 g/mol
- Seq:9220
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
:
:235.09 g/mol
:GHS07
:Refrigerator
:408502-23-8
:C₁₂H₁₈BNO₃
:MFCD07368867