
6-Methyleneandrost-4-ene-3,17-dione ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:19457-55-7
:
Formula:C₂₀H₂₆O₂
MDL Number:MFCD09038727
:298.43 g/mol
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+390 to +430 deg (C=1, CHCL3)
Specification Test Results
Frequently Bought Together