4'-Methoxy-N,N-di-p-tolyl-[1,1'-biphenyl]-4-amine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:129119-41-1
:
Formula:C₂₇H₂₅NO
:379.50 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
Frequently Bought Together