

Mitoxantrone dihydrochloride ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:70476-82-3
:
Formula:C₂₂H₃₀Cl₂N₄O₆
MDL Number:MFCD00242943
:517.404 g/mol
Merck Index:14,06217
Storage Temperature:Refrigerator
EINECS:274-619-1
UN:2811
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Dark green to Dark purple to Black powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Frequently Bought Together