
Methyl-2-bromo-5-fluorobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6942-39-8
:
Formula:C₈H₆BrFO₂
MDL Number:MFCD00086167
:233.04 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Acid Value:max. 2.0
Specification Test Results
Frequently Bought Together